Skip to Content
Merck
All Photos(1)

Key Documents

52284

Millipore

Lactose Fermentation Membrane

white colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
€26.00

€26.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
25 DISCS
€26.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

€26.00


Please contact Customer Service for Availability

Request more information

membrane size

70 mm

Quality Level

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
bacteria
coliforms

General description

Lactose Fermentation Membranes are substrate-carrying ID membranes used for rapid identification and differentiation of lactose fermenting organisms (coliforms) through characteristic color reactions. Microorganisms can only ferment certain carbohydrates, depending on the enzymes they possess to break down carbohydrates. The membranes contain lactose as substrates and a pH indicator, which serve as the basis for microbe differentiation by color. The test is specific for lactose fermenting organisms, which will exhibit yellow color.

Application

Lactose Fermentation Membranes are recommended to detect lactose fermenting bacteria (coliforms) in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Citrobacter freundii (8090) - PositiveEnterobacter aerogenes (13048) - PositiveEscherichia coli (25922) - PositiveKlebsiella pneumoniae (13883) - PositiveProteus vulgaris (13315)- NegativeSalmonella typhimurium (14028) - NegativeSalmonella typhi (6539) - NegativeSerratia marcescens (8100) - NegativeShigella flexneri (12022) - Negative

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service