Skip to Content
Merck
All Photos(2)

Key Documents

51691

Supelco

1,2,5,6,9,10-Hexabromocyclododecane

analytical standard

Synonym(s):

HBCD, HBCDD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Br6
CAS Number:
Molecular Weight:
641.70
Beilstein:
1911324
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (sum of isomers, HPLC)
95.0-105.0% (wt%, argentometric after oxygen combustion)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: slightly soluble

format

neat

SMILES string

BrC1CCC(Br)C(Br)CCC(Br)C(Br)CCC1Br

InChI

1S/C12H18Br6/c13-7-1-2-8(14)10(16)5-6-12(18)11(17)4-3-9(7)15/h7-12H,1-6H2

InChI key

DEIGXXQKDWULML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Suzuki et al.
The Journal of sports medicine and physical fitness, 54(5), 622-630 (2014-10-02)
Aim of the present study was to compare the effects of highly branched cyclic dextrin (HBCD) drink with a glucose-based control drink on immunoendocrine responses to endurance exercise. Using a randomized, double-blind placebo-controlled cross-over design, seven male triathletes participated in
Reza Farmahin et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 133, 110262-110262 (2018-12-31)
Hexabromocyclododecane (HBCD) is a brominated flame retardant found in the environment and human tissues. The toxicological effects of HBCD exposure are not clearly understood. We employed whole-genome RNA-sequencing on liver samples from male and female Fischer rats exposed to 0
A Polder et al.
The Science of the total environment, 488-489, 252-260 (2014-05-20)
In Tanzania fish is one of the most important protein sources for the rapidly increasing population. Wild fish is threatened by overfishing and pollution from agriculture, industries, mining, household effluents and vector control. To monitor possible implications for public health
Li Zhang et al.
Journal of chromatography. A, 1386, 22-30 (2015-02-24)
A new analytical method was established and validated for the analysis of 19 substances of very high concern (SVHCs) in textiles, including phthalic acid esters (PAEs), organotins (OTs), perfluorochemicals (PFCs) and flame retardants (FRs). After ultrasonic extraction in methanol, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service