51535
Methyl heneicosanoate
analytical standard
Synonym(s):
Heneicosanoic acid methyl ester
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
bp
207 °C/4 mmHg (lit.)
mp
48-50 °C (lit.)
48-50 °C
format
neat
functional group
ester
shipped in
ambient
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCCCCCC(=O)OC
InChI
1S/C22H44O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-2/h3-21H2,1-2H3
InChI key
AJRICDSAJQHDSD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service