Skip to Content
Merck
All Photos(1)

Key Documents

50953

Supelco

Bisphenol G

analytical standard

Synonym(s):

2,2-Bis(4-hydroxy-3-isopropylphenyl)propan, 4,4′-(1-Methylethylidene)bis[2-(1-methylethyl)phenol], 4,4′-Isopropylidenebis(2-isopropylphenol), NSC 21526

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Beilstein:
2383602
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(C=C1)=C(C(C)C)C=C1C(C)(C)C2=CC=C(O)C(C(C)C)=C2

InChI

1S/C21H28O2/c1-13(2)17-11-15(7-9-19(17)22)21(5,6)16-8-10-20(23)18(12-16)14(3)4/h7-14,22-23H,1-6H3

InChI key

IJWIRZQYWANBMP-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antía Lestido Cardama et al.
Polymers, 11(12) (2019-12-19)
The coatings used in cans can release complex chemical mixtures into foodstuffs. Therefore, it is important to develop analytical methods for the identification of these potential migrant compounds in packaged food to guarantee the compliance with European food packaging legislation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service