Skip to Content
Merck
All Photos(1)

Key Documents

47426

Sigma-Aldrich

Fmoc-Phe(3,4-Cl2)-OH

≥96.0% (HPLC)

Synonym(s):

Fmoc-3,4-dichloro-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H19Cl2NO4
CAS Number:
Molecular Weight:
456.32
Beilstein:
9016646
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥96.0% (HPLC)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccc(Cl)c(Cl)c1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H19Cl2NO4/c25-20-10-9-14(11-21(20)26)12-22(23(28)29)27-24(30)31-13-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-11,19,22H,12-13H2,(H,27,30)(H,28,29)/t22-/m0/s1

InChI key

QNVHCYWPXIGFGN-QFIPXVFZSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerardo Alfonso Anguiano-Vega et al.
Scientific reports, 10(1), 17584-17584 (2020-10-18)
This study identified and determined organochloride pesticide (OCs) concentrations in hair samples from children at two elementary schools: one exposed to fumigations in agricultural fields, the other unexposed. Three concentrations of OCs levels in the hair were compared (high, medium
Andrea Escamilla-López et al.
Archives of environmental contamination and toxicology, 78(2), 303-309 (2019-11-09)
Organochlorine pesticides (OCPs) have been used for many decades, both for the control of pests in agriculture and for the control of vectors of human and animal diseases. Several recent studies have reported significant concentrations of these compounds in multiple
Gabriela García-Besné et al.
Marine pollution bulletin, 91(1), 139-148 (2015-01-01)
Organochlorine pesticides and PCB (POPs) concentrations were determined in the blood and eggs of green and hawksbill turtles. We compared concentrations between species, analyzed the relationship between turtle size and the POPs concentrations and the relationship between the concentrations in
Patricia I Salvarani et al.
Archives of environmental contamination and toxicology, 76(3), 425-434 (2019-01-03)
The investigation of organochlorine pesticides (OCPs) levels in sea turtles is an important issue in conservation research, due to the harmful effects of these chemicals. In the present study, OCPs concentrations were determined in the eggs of two sea turtle
Winnette A Collimore et al.
Environmental monitoring and assessment, 192(2), 128-128 (2020-01-23)
A newly modified QuEChERS method was developed, validated, and tested in-house for the multiclass, multiresidue determination of organophosphate (OPP) and organochlorine pesticide (OCP) residues in fruits and vegetables. The method incorporated a solvent extraction with acetonitrile followed by partitioning with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service