Skip to Content
Merck
All Photos(1)

Key Documents

46408

Supelco

Stearyl stearate

analytical standard

Synonym(s):

Stearic acid stearyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€128.00

€128.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
€128.00

About This Item

Linear Formula:
CH3(CH2)16COO(CH2)17CH3
CAS Number:
Molecular Weight:
536.96
Beilstein:
1807499
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€128.00


Available to ship on03 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

ester

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C36H72O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-38-36(37)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-35H2,1-2H3

InChI key

NKBWPOSQERPBFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Stearyl stearate is the ester of stearic acid and 1-octadecanol.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Stearyl stearate may be used as a model lipid to stain contact lenses in evaluating the lipid-degrading ability of the cholesterol esterase enzyme produced from Pseudomonas aeruginosa.[2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and characterization of a novel cholesterol esterase from Pseudomonas aeruginosa, with its application to cleaning lipid-stained contact lenses.
Sugihara A.
Bioscience, Biotechnology, and Biochemistry, 66(11), 2347-2355 (2002)
Derivatisation using m-(trifluoromethyl) phenyltrimethylammonium hydroxide of organic materials in artworks for analysis by gas chromatography?mass spectrometry: unusual reaction products with alcohols.
Sutherland K.
Journal of Chromatography A, 1149(1), 30-37 (2007)
Dorothy M Duffy et al.
Langmuir : the ACS journal of surfaces and colloids, 20(18), 7630-7636 (2004-08-25)
Living organisms can control the size, shape, and structure of minerals. Attempts to reproduce this biological control in the laboratory often use Langmuir monolayers of long-chain carboxylic acids. We use large-scale molecular dynamics simulations to calculate the interfacial energies of
K Maeda et al.
Journal of chromatography, 221(2), 199-204 (1980-12-12)
A gas chromatographic--mass spectrometric analysis was used to separate and identify abnormal compounds in the nail of psoriatic patients. The nail was extracted with heated ethanol, and the extract was analyzed with and without trimethylsilylation. Tetradecanoic acid octadecyl ester, hexadecanoic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service