Skip to Content
Merck
All Photos(1)

Key Documents

46336

Supelco

Cinidon-ethyl

PESTANAL®, analytical standard

Synonym(s):

cis-Ethyl 2-chloro-3-[2-chloro-5-(1,3,4,5,6,7-hexahydro-1,3-dioxo-2H-isoindol-2-yl)phenyl]-2-propenoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€97.00

€97.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€97.00

About This Item

Empirical Formula (Hill Notation):
C19H17Cl2NO4
CAS Number:
Molecular Weight:
394.25
Beilstein:
8446817
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€97.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)\C(Cl)=C\c1cc(ccc1Cl)N2C(=O)C3=C(CCCC3)C2=O

InChI

1S/C19H17Cl2NO4/c1-2-26-19(25)16(21)10-11-9-12(7-8-15(11)20)22-17(23)13-5-3-4-6-14(13)18(22)24/h7-10H,2-6H2,1H3/b16-10-

InChI key

NNKKTZOEKDFTBU-YBEGLDIGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cinidon-ethyl is a herbicide used to control broadleaf weeds in cereals.[1]

Application

Cinidon-ethyl may be used as a reference standard for the analysis of cinidon-ethyl pesticide residue in fruits and vegetables using QuEChERS method followed by liquid chromatography coupled with mass spectrometry (LC–MS/MS).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of Ethylene in the Induction of Cotton Defoliation by the Isoindoldione Cinidon-ethyl.
Grossmann K.
Pesticide Biochemistry and Physiology, 68(1), 42-48 (2000)
QuEChERS analytical method for determination of 93 pesticide residues in apples and potatoes using LC--MS/MS.
Attallah E R, et al.
Journal of Applied Sciences Research, 8, 5690-5697 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service