Skip to Content
Merck
All Photos(1)

Key Documents

45996

Supelco

1,2,4-Trimethylbenzene

analytical standard

Synonym(s):

Pseudocumene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
1903005
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39011403
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.1 (vs air)

vapor pressure

4.5 mmHg ( 37.7 °C)

autoignition temp.

959 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.504 (lit.)

bp

168 °C (lit.)

mp

−44 °C (lit.)

density

0.876 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1ccc(C)c(C)c1

InChI

1S/C9H12/c1-7-4-5-8(2)9(3)6-7/h4-6H,1-3H3

InChI key

GWHJZXXIDMPWGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,4-Trimethylbenzene may be used as an analytical reference standard for the quantification of the analyte in environmental samples using gas chromatography coupled to mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase microextraction (SPME) for rapid field sampling and analysis by gas chromatography-mass spectrometry (GC-MS)
Hook.LG, et al.
TrAC, Trends in Analytical Chemistry, 21(8), 534-543 (2002)
Fei Li et al.
Indian journal of dermatology, 65(2), 92-96 (2020-03-18)
Common indoor pollutants, as fine particulate matter (PM2.5), can damage people's health and cause skin allergies. However, it remains unknown which common pollutants can lead to allergy, such as, in children atopic dermatitis, and what is the key molecule. This
Birger Bohn et al.
Physical chemistry chemical physics : PCCP, 14(40), 13933-13948 (2012-09-13)
The reversible gas-phase addition of OH radicals to the trimethylbenzenes was investigated in pulsed experiments utilizing VUV flash-photolysis resonance-fluorescence of H(2)O in the temperature range of 275-340 K. Triexponential OH decays were observed in the presence of the trimethylbenzenes, indicating
Hans H Richnow et al.
Journal of contaminant hydrology, 64(1-2), 59-72 (2003-05-15)
Concentrations and isotopic compositions (13C/12C) of aromatic hydrocarbons were determined in eight samples obtained from the strongly anoxic part of the leachate plume downgradient from the Vejen Landfill (Denmark), where methanogenic, sulfate-reducing and iron-reducing conditions were observed. Despite the heterogeneous
C M Kao et al.
Water research, 35(8), 1951-1960 (2001-05-08)
Natural bioremediation is believed to be the major processes that account for both containment of the petroleum-hydrocarbon plume and reduction of the contaminant concentrations. In this study, the feasibility of applying the microbial enumeration technique was assessed for natural biodegradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service