Skip to Content
Merck
All Photos(1)

Key Documents

45925

Supelco

1-Chloro-4-nitrobenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
508691
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.4 (vs air)

vapor pressure

0.09 mmHg ( 25 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

242 °C (lit.)

mp

80-83 °C (lit.)

density

1.298 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[O-][N+](=O)c1ccc(Cl)cc1

InChI

1S/C6H4ClNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H

InChI key

CZGCEKJOLUNIFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Muta. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Miaomiao Ye et al.
Journal of hazardous materials, 184(1-3), 612-619 (2010-09-22)
TiO(2) hollow microspheres were synthesized by a simple hydrothermal method followed by calcination at different temperatures ranging from 400 to 800°C. The prepared samples were characterized by XRD, SEM, TEM, SAED, HRTEM, N(2) adsorption, and UV-vis spectroscopy. The photocatalytic activities
Liang Zhu et al.
Water research, 46(19), 6291-6299 (2012-10-17)
Using a combined zero-valent iron (ZVI) and anaerobic sludge system as the platform, the hydrogen utilization and reductive mechanism of p-chloronitrobenzene (p-ClNB) were investigated in the study. Results showed that the corrosion of ZVI could be used as a source
Yun Zhang et al.
Biodegradation, 20(1), 55-66 (2008-05-30)
Comamonas sp. strain CNB-1 can utilize 4-chloronitrobenzene (4CNB) as sole carbon and nitrogen source for growth. Previous studies were focused on 4CNB degradative pathway and have showed that CNB-1 contained a plasmid pCNB1 harboring the genes (cnbABCaCbDEFGH, cnbZ) for the
Min Mo et al.
Chemical communications (Cambridge, England), 46(13), 2268-2270 (2010-03-18)
High quality noncrystalline NiPB nanotubes were synthesized and exhibited high efficiency for the catalytic hydrogenation of p-chloronitrobenzene due to the characteristic confinement effect of the nanotubes.
Chen Le et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 64(10), 2126-2131 (2011-11-23)
In this study, zero-valent iron (ZVI) was used to pretreat para-chloronitrobenzene (p-CNB), and the major product was para-chloroaniline (p-CAN). By adding H(2)O(2) directly, further p-CAN degradation can be attributed to Fenton oxidation because ferrous ions (Fe(2+)) released during the ZVI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service