Skip to Content
Merck
All Photos(1)

Documents

45913

Supelco

Trifluralin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16F3N3O4
CAS Number:
Molecular Weight:
335.28
Beilstein:
1893555
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CCCN(CCC)c1c(cc(cc1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

InChI

1S/C13H16F3N3O4/c1-3-5-17(6-4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8H,3-6H2,1-2H3

InChI key

ZSDSQXJSNMTJDA-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V K Shormanov et al.
Sudebno-meditsinskaia ekspertiza, 53(5), 31-33 (2011-01-27)
Results of extraction of trifluralin from an aqueous acetonitrile solution using various organic solvents are presented. The degree of extraction was shown to depend on the nature of extractant and the water/acetonitrile ratio. An optimal electrolyte and the degree of
Guillaume Salquèbre et al.
Analytica chimica acta, 710, 65-74 (2011-11-30)
A method for the simultaneous detection and quantification of 22 pesticides from different chemical classes was developed using solid-phase microextraction (SPME) and gas chromatography tandem (triple quadrupole) mass spectrometry. Pesticides were extracted from 50mg of pulverized hair with acetonitrile. The
Perihan Binnur Kurt-Karakus et al.
Environmental toxicology and chemistry, 30(7), 1539-1548 (2011-04-08)
Concentrations of current-use pesticides (CUPs) in water, zooplankton, precipitation, and air samples as well as stereoisomer fractions (SF; herbicidally active/total stereoisomers) of metolachlor were determined in water samples collected from 10 remote inland lakes in Ontario, Canada, between 2003 and
Belma Aslim et al.
Journal of environmental biology, 30(3), 381-384 (2010-02-03)
The herbicides most commonly used in Turkey are trifluralin (alpha,alpha,alpha-trifluoro-2,6-dinitro-N,N-dipropyl-p-toluidine), 2,4-D (2,4-dichlorophenoxyacetic acid), and linuron [3-(3,4-dichlorophenyl)-N-methoxy-N-methylurea]. The effects of these three herbicides on the growth of 10 threatened aquatic cyanobacterial isolates were tested by using 9-day exposure experiments at concentrations
[The development of a method for chemico-toxicological analysis of trifluralin].
V K Shormanov et al.
Sudebno-meditsinskaia ekspertiza, 54(3), 50-52 (2011-08-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service