Skip to Content
Merck
All Photos(1)

Key Documents

45816

Supelco

Bromfenvinphos-ethyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14BrCl2O4P
CAS Number:
Molecular Weight:
404.02
Beilstein:
6069447
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

description

mixture of cis and trans isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)O\C(=C\Br)c1ccc(Cl)cc1Cl

InChI

1S/C12H14BrCl2O4P/c1-3-17-20(16,18-4-2)19-12(8-13)10-6-5-9(14)7-11(10)15/h5-8H,3-4H2,1-2H3/b12-8+

InChI key

ORDKAVSHIKNMAN-XYOKQWHBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Ultrastructure of rat liver cells in acute and shortened chronic poisoning with bromphenvinphos (author's transl)].
J Ochyński
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina, 33, 35-40 (1978-01-01)
[In vitro studies of the effect of organophosphate compounds on the surface structure of monkey kidney cells].
T Skwarek et al.
Medycyna doswiadczalna i mikrobiologia, 36(2), 129-133 (1984-01-01)
K I Zaleska-Freljan et al.
Polish journal of pharmacology and pharmacy, 35(3), 185-193 (1983-05-01)
Bromfenvinphos (0.0-diethyl-0-1-(2.4-dichlorphenyl)-2-bromvinyl phosphate), BrV, 12.33 mg/kg/day, alone and in combination with methoxychlor ( (1,1,1-trichlor-2.2-)4-methoxyphenyl ethane), MeOCl, 24.66 mg/kg/day, were administered to laboratory mice in the olive-oil suspension through the stomach gauge once a day for 6 weeks. The histological changes
Influence of chlorfenvinphos and ipofos on oxido-reducing processes in the rat brain mitochondria during development.
M Skonieczna et al.
Neuropatologia polska, 19(2), 197-208 (1981-04-01)
[Effectiveness of bromfenvinphos and photostable pyrethrin mixtures in the control of sanitary pests].
J Krocznński et al.
Wiadomosci parazytologiczne, 32(4-6), 527-530 (1986-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service