Skip to Content
Merck
All Photos(1)

Key Documents

45721

Supelco

Chlorflurenol

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-9-hydroxyflurene-9-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€107.00

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€107.00

About This Item

Empirical Formula (Hill Notation):
C14H9ClO3
CAS Number:
Molecular Weight:
260.67
Beilstein:
3344166
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)C1(O)c2ccccc2-c3ccc(Cl)cc13

InChI

1S/C14H9ClO3/c15-8-5-6-10-9-3-1-2-4-11(9)14(18,13(16)17)12(10)7-8/h1-7,18H,(H,16,17)

InChI key

SVOAUHHKPGKPQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatih Kapukıran et al.
Bulletin of environmental contamination and toxicology, 102(6), 848-853 (2019-04-17)
In this study, the detection power of a gas chromatography mass spectrometer (GC-MS) for procymidone and chlorflurenol was significantly enhanced using switchable solvent liquid phase microextraction (SS-LPME) as a preconcentration tool. This was achieved by a comprehensive optimization of significant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service