Skip to Content
Merck
All Photos(2)

Documents

45664

Supelco

Sulfotep

PESTANAL®, analytical standard

Synonym(s):

O,O,O′,O′-Tetraethyl dithiopyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20O5P2S2
CAS Number:
Molecular Weight:
322.32
Beilstein:
1714019
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)OP(=S)(OCC)OCC

InChI

1S/C8H20O5P2S2/c1-5-9-14(16,10-6-2)13-15(17,11-7-3)12-8-4/h5-8H2,1-4H3

InChI key

XIUROWKZWPIAIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfotep is a non-systemic, aliphatic organophosphate insecticide, fumigant and acaricide, which may cause unanticipated health and ecological problems. It acts as a cholinesterase inhibitor, and is widely used in controlling aphids, spider mites, thrips and whiteflies on glasshouse crops, mushrooms and ornamentals.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfotep may be used as a reference standard for the determination of the analyte in liquid pesticide formulations using gas chromatography with flame ionization detection (GC-FID).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Simultaneous gas chromatographic determination of chlorpyrifos and its impurity sulfotep in liquid pesticide formulations.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 51(11), 736-741 (2016)
The Pesticide Encyclopedia (2014)
MMWR. Morbidity and mortality weekly report, 45(36), 780-782 (1996-09-13)
Pesticide fumigants that eradicate pests but do not damage flowers or foliage can be used to protect market-ready florals. During November 1995, a pesticide applicator worker in Texas became ill during fumigation despite wearing the personal protective equipment (PPE) recommended
Determination of chlorpyrifos and its major breakdown products in technical formulations.
W J Allender et al.
Bulletin of environmental contamination and toxicology, 46(2), 313-319 (1991-02-01)
Lili Pan et al.
Environmental science and pollution research international, 25(1), 4-11 (2016-10-01)
Organophosphorus pesticides (OPPs) are used worldwide and pose great risks to human health. However, information on their presence in agricultural soils at regional scale and the associated risks is limited. In this study, an extensive investigation on agricultural soils was

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service