Skip to Content
Merck
All Photos(2)

Key Documents

45660

Supelco

Simetryn

PESTANAL®, analytical standard

Synonym(s):

2,4-Bis(ethylamino)-6-methylthio-1,3,5-triazine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€21.20

€21.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€21.20

About This Item

Empirical Formula (Hill Notation):
C8H15N5S
CAS Number:
Molecular Weight:
213.30
Beilstein:
11728
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€21.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NCC)nc(SC)n1

InChI

1S/C8H15N5S/c1-4-9-6-11-7(10-5-2)13-8(12-6)14-3/h4-5H2,1-3H3,(H2,9,10,11,12,13)

InChI key

MGLWZSOBALDPEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas S Machado et al.
Journal of chromatography. A, 1651, 462260-462260 (2021-06-06)
Monolithic polymers are described as continuous and highly porous materials. They have been gaining popularity as an effective extracting phase for some sample preparation methods, due to their variety of functionalities, such as wide pH range tolerance, good permeability, and
Russel C G Chidya et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 53(9), 602-614 (2018-07-24)
This study investigated spatio-temporal variations of selected pesticide residues in the Kurose River in Higashi-Hiroshima city (Hiroshima Prefecture), Japan. Water samples were collected from the river at seven sites every month for 1 year (March 2016 to February 2017). Pesticide residues
Suqin Wu et al.
Journal of chromatography. A, 1218(10), 1340-1346 (2011-01-29)
The homogeneous molecularly imprinted microspheres (MIMs) based on a biologically inspired hydrogen-bond array were prepared using allobarbital as the novel functional monomer and divinylbenzene as the cross-linker. The host-guest binding characteristics were examined by molecular simulation and infrared spectroscopy. The
G J Hughes et al.
The Biochemical journal, 199(1), 61-67 (1981-10-01)
Native rat haemoglobins were found to bind simetryn sulphoxide to an extent 40-fold greater than human haemoglobin. This specific behaviour was studied by using only high-pressure ('performance') liquid chromatography for the preparative separation of globin chains and the isolation of
A Numabe et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 53(2), 139-146 (2006-04-06)
The runoff characteristics of pesticides from paddy fields to rural rivers were investigated over a period of three years in Hokkaido Prefecture, Japan. High pesticide concentrations were usually observed in rivers during pesticide application periods. In one year, the growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service