Skip to Content
Merck
All Photos(1)

Key Documents

45659

Supelco

Methyl cis-13-docosenoate

analytical standard

Synonym(s):

Methyl erucate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€73.20
5 ML
€221.00

€73.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€73.20
5 ML
€221.00

About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)11COOCH3
CAS Number:
Molecular Weight:
352.59
Beilstein:
1728408
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€73.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.456

bp

172-175 °C/0.2 mmHg (lit.)

density

0.870 g/mL at 20 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCCCCC(=O)OC

InChI

1S/C23H44O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h10-11H,3-9,12-22H2,1-2H3/b11-10-

InChI key

ZYNDJIBBPLNPOW-KHPPLWFESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayesha Khalid et al.
Scientific reports, 8(1), 14883-14883 (2018-10-07)
Acute Leukaemia (AL) is a neoplasm of WBCs (white blood cells). Being an important class of metabolites, alteration in free fatty acids (FFAs) levels play a key role in cancer development and progression. As they involve in cell signaling, maintain
Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and

Questions

  1. what concentration is the product code 45659-1ML-F?

    1 answer
    1. The concentration for product 45659-1ML-F will be lot specific depending on the purity assay using the formula: Density X 1000 X Concentration/ MW = Molarity (M). For example, lot BCCH6579 has a purity result of 99.4% which gives 0.870 g/mL X 1000 X 0.994/ 352.59 g/mol=2.45 M.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service