Skip to Content
Merck
All Photos(1)

Key Documents

45646

Supelco

Pyracarbolid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€67.80

€67.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€67.80

About This Item

Empirical Formula (Hill Notation):
C13H15NO2
CAS Number:
Molecular Weight:
217.26
Beilstein:
1427914
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€67.80


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(NC=1C=CC=CC1)C2=C(OCCC2)C

InChI

1S/C13H15NO2/c1-10-9-11(7-8-16-10)13(15)14-12-5-3-2-4-6-12/h2-6,9,11H,7-8H2,1H3,(H,14,15)

InChI key

RVUAOAREEJAWAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyracarbolid is an anilide compound, used as a systemic foliar fungicide which is found to be effective against coffee rust.[1]

Application

Pyracarbolid may be used as an analytical reference standard for the quantification of the analyte in fruit juice samples using liquid chromatography and gas chromatography coupled to tandem mass spectrometry.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coffee Rust: Epidemiology, Resistance, and Management (1989)
Finding of pesticides in fashionable fruit juices by LC-MS/MS and GC-MS/MS.
Tran K, et al.
Food Chemistry, 134(4), 2398-2405 (2012)
Kaan Georg Kutlucinar et al.
Electrophoresis, 42(4), 490-500 (2020-12-18)
Nontargeted analysis of water samples using liquid chromatography combined with high-resolution mass spectrometers is an emerging approach for surface water monitoring and evaluation of water treatment processes. In this study, sample preconcentration via direct, large volume injection with 500 μL and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service