Skip to Content
Merck
All Photos(1)

Key Documents

45475

Supelco

Ethiofencarb

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2S
CAS Number:
Molecular Weight:
225.31
Beilstein:
2973224
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCSCc1ccccc1OC(=O)NC

InChI

1S/C11H15NO2S/c1-3-15-8-9-6-4-5-7-10(9)14-11(13)12-2/h4-7H,3,8H2,1-2H3,(H,12,13)

InChI key

HEZNVIYQEUHLNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pérez Clavijo et al.
Journal of chromatography. A, 740(1), 146-150 (1996-07-26)
A method for monitoring pesticides in apple samples, including Soxhlet extraction, an evaporation step and capillary gas chromatography with nitrogen-phosphorus detection, is applied to a decay study of the carbamate pesticide ethiofencarb. The evaporation step is carried out by a
[Polynuclear genital cells in rat spermatogenesis in the normal state and after exposure to xenobiotics].
Iu V Ivanov
Gigiena truda i professional'nye zabolevaniia, (3)(3), 54-55 (1989-01-01)
[Hygienic characteristics of the pesticide croneton].
V N Karpenko et al.
Gigiena i sanitariia, (10)(10), 23-26 (1984-10-01)
H Tanaka et al.
Journal of toxicology and environmental health, 47(5), 493-498 (1996-04-05)
Ethiofencarb and pirimicarb, widely used insecticides, were investigated for hepatocarcinogenesis-modifying effects using a medium-term bioassay system for carcinogens. F344 male rats were given a single intraperitoneal injection of diethylnitrosamine (DEN, 200 mg/kg) and then, starting 2 wk later, received ethiofencarb
[Determination of croneton in the air].
G A Khokhol'kova et al.
Gigiena i sanitariia, (12)(12), 75-76 (1982-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service