Skip to Content
Merck
All Photos(1)

Key Documents

44967

Supelco

Cerium(III) Ionophore

Selectophore, function tested

Synonym(s):

2-Aminobenzothiazole, 2-Benzothiazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2S
CAS Number:
Molecular Weight:
150.20
Beilstein:
116315
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

form

solid

quality

function tested

mp

126-129 °C (lit.)

SMILES string

Nc1nc2ccccc2s1

InChI

1S/C7H6N2S/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H2,8,9)

InChI key

UHGULLIUJBCTEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Aminobenzothiazole is a benzothiazole derivative. These derivatives are heterocycles consisting of thiazole, sulphur and nitrogen moieties.[1]
Visit our Sensor Applications portal to learn more.

Application

It was used in the synthesis of 2-(2′ -benzothiazolylazo)-5-(N,N-diethyl)aminophenol (BDAP) during the determination of copper and nickel in food samples using cloud-point extraction method.[2]

Other Notes

Preparation of a cerium(III)-selective membrane electrode[3]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of 2-aminobenzothiazole derivatives.
Srivastava, S. D., and J. P. Sen.
Indian J. Chem. B, 47.10, 1583-1583 (2008)
A new cerium (III)-selective membrane electrode based on 2-aminobenzothiazole
M. Akhond et al.
Sensors and Actuators B, Chemical, 99, 410-410 (2004)
Valfredo Azevedo Lemos et al.
Journal of hazardous materials, 159(2-3), 245-251 (2008-03-21)
A new, simple and versatile cloud-point extraction (CPE) methodology has been developed for the separation and preconcentration of copper and nickel. The metals in the initial aqueous solution were complexed with 2-(2'-benzothiazolylazo)-5-(N,N-diethyl)aminophenol (BDAP) and Triton X-114 was added as surfactant.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service