Skip to Content
Merck
All Photos(1)

Key Documents

44579

Sigma-Aldrich

Drierite

drying agent

Synonym(s):

Calcium sulfate, Gypsum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4
CAS Number:
Molecular Weight:
136.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

drying agent

particle size

10-20 mesh

SMILES string

[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

OSGAYBCDTDRGGQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ontogeny of personality in the wild guinea pig.
Guenther A, et al.
Animal behaviour, 90, 131-139 (2014)
The analysis of carbon monoxide in urban air at the ppm level, and the normal carbon monoxide value.
L Dubois et al.
Journal of the Air Pollution Control Association, 16(3), 135-139 (1966-03-01)
Enthalpies of vaporization of organic compounds.
Howard PB and Wadso I.
Acta Chemica Scandinavica, 24, 145-149 (1970)
Nathaniel Huebsch et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), 9762-9767 (2014-06-26)
Biological systems are exquisitely sensitive to the location and timing of physiologic cues and drugs. This spatiotemporal sensitivity presents opportunities for developing new therapeutic approaches. Polymer-based delivery systems are used extensively for attaining localized, sustained release of bioactive molecules. However
Respirometry: anhydrous drierite equilibrates with carbon dioxide and increases washout times.
White CR, et al.
Physiological and Biochemical Zoology : Pbz, 79(5), 977-988 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service