Skip to Content
Merck
All Photos(1)

Key Documents

443468

Sigma-Aldrich

2-Butanone

ACS reagent, ≥99.0%

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
EC Number:
MDL number:
UNSPSC Code:
12190000

grade

ACS reagent

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

Assay

≥99.0%

autoignition temp.

960 °F

expl. lim.

10.1 %

impurities

≤0.0005 meq/g Titr. acid
≤0.20% water

evapn. residue

≤0.0025%

color

APHA: ≤15

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Butanone (Methyl ethyl ketone, MEK), an aliphatic ketone, is considered as aprotic and protophilic solvent. The solvent composition plays an important role in the rate of liquid-phase catalytic hydrogenation of MEK to 2-butanol. The rate constant for the reaction of hydroxyl radicals with MEK has been determined and expressed in Arrhenius form as ΚC2H5COCH3+OH = 6.35*1013 exp(-2270/T) cm3mol-1s-1.

Application

2-Butanone may be used as an oxidizing agent for the conversion of primary alcohols to the corresponding aldehydes via Oppenauer oxidation in the presence of an iridium complex.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

High-Temperature Measurements of the Reactions of OH with a Series of Ketones: Acetone, 2-Butanone, 3-Pentanone, and 2-Pentanone.
Lam KY, et al.
The Journal of Physical Chemistry A, 116(23), 5549-5559 (2012)
Densities and viscosities of binary liquid mixtures of 2-butanone with branched alcohols at (293.15 to 313.15)K.
Lomte SB, et al.
Journal of Chemical and Engineering Data, 54(1), 127-130 (2008)
Solvent effects in the hydrogenation of 2-butanone.
Akpa BS, et al.
J. Catal., 289, 30-41 (2012)
Iridium-catalyzed Oppenauer oxidations of primary alcohols using acetone or 2-butanone as oxidant.
Suzuki T, et al.
The Journal of Organic Chemistry, 68(4), 1601-1602 (2003)
Shane Avison et al.
Journal of agricultural and food chemistry, 63(28), 6313-6318 (2015-06-23)
A new methodology is presented to measure water-air partition coefficients (Henry's constants) of volatiles, using APCI-MS. Significant advantages over other Henry's constant determination methods include the short measurement and sample preparation time and the possibility for simultaneous measurement of multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service