43856
Alkaline Peptone Water (ISO)
suitable for microbiology
Synonym(s):
Peptone Water, alkaline (ISO)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
shelf life
limited shelf life, expiry date on the label
composition
peptic digest of animal tissue, 20 g/L
sodium chloride, 30 g/L
final pH
8.6±0.2 (25 °C)
application(s)
microbiology
suitability
nonselective for Vibrio spp.
Application
Alkaline Peptone Water is used for detection of Vibrio species. It is also recommended by ISO Committee under the specification ISO 8914:1990.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service