Skip to Content
Merck
All Photos(1)

Key Documents

433934

Supelco

Amberlite HPR4100 Ion Exchange Resin

chloride form, 30-35 mesh, anionic

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€255.00

€255.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€255.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

€255.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

AmberLite HPR4100 Cl Anion Exchange Resin,

description

formerly DowexA2 chloride

Quality Level

quality

monodispersed

moisture

45-54%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

dimethylethanolamine

particle size

27-34 mesh
440-560 μm

capacity

1.2 meq/mL by wetted bed volume

separation technique

anion exchange

General description

AmberLiteHPR4100 Cl has dimethylethanolamine as functional group.[1]

Application

AmberLite HPR4100 Cl was used in structural characterization of Fe in dilute natural waters.[1]
Strongly basic (type II) anion exchange resin well suited for water with high concentration of mineral acids (chlorides, sulfates) and low concentration of silica and CO2 (<25%). This resin has excellent efficiency for general demineralization, and meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

Strongly basic anion

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anneli Sundman et al.
Environmental science & technology, 47(15), 8557-8564 (2013-07-03)
The properties of iron (Fe) complexes and compounds in the environment influence several central processes, e.g., iron uptake, adsorption/desorption of contaminants and nutrients, and redox transformations, as well as the fate of of natural organic matter (NOM). It is thus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service