Skip to Content
Merck
All Photos(1)

Key Documents

43165

Sigma-Aldrich

trans-2,3-Diphenyl-1-phthalimidoaziridine

purum, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N2O2
CAS Number:
Molecular Weight:
340.37
Beilstein:
1395096
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (HPLC)

mp

174-176 °C

SMILES string

O=C1N(N2[C@@H]([C@H]2c3ccccc3)c4ccccc4)C(=O)c5ccccc15

Other Notes

Stable precursor for 1-amino-trans-2,3-diphenylaziridine[1]; The free hydrazine is a reagent for α,β-epoxyketone#8594;alkynone fragmentation[2][3]; Reagent used for the key cyclization step in the synthesis of a isoindolobenzazepine[4]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Felix et al.
Helvetica Chimica Acta, 55, 1276-1276 (1972)
F.G. Fang et al.
Tetrahedron Letters, 30, 2747-2747 (1989)
Organic Syntheses, 55, 52-52 (1976)
R.K. Muller et al.
Organic Syntheses, 55, 114-114 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service