Skip to Content
Merck
All Photos(1)

Key Documents

43084

Supelco

Ginsenoside Rh3

analytical standard

Synonym(s):

(3β, 12β,20Z)-12-Hydroxydammara-20(22),24-dien-3-yl-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60O7
CAS Number:
Molecular Weight:
604.86
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

InChI

1S/C36H60O7/c1-20(2)10-9-11-21(3)22-12-16-36(8)28(22)23(38)18-26-34(6)15-14-27(33(4,5)25(34)13-17-35(26,36)7)43-32-31(41)30(40)29(39)24(19-37)42-32/h10-11,22-32,37-41H,9,12-19H2,1-8H3/b21-11-/t22-,23+,24-,25?,26?,27?,28+,29-,30+,31-,32+,34+,35-,36+/m1/s1

InChI key

PHLXREOMFNVWOH-WJJFPOJFSA-N

General description

Ginsenoside Rh3 is a metabolite of ginsenoside Rg5, isolated from Panax ginseng.

Application

Ginsenoside Rh3 may be used as a reference standard in the determination of ginsenoside Rg5 in Korean red ginseng using high performance liquid chromatography coupled with evaporative light scattering detection (HPLC-ELSD).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ginsenoside Rg5 improves cognitive dysfunction and beta-amyloid deposition in STZ-induced memory impaired rats via attenuating neuroinflammatory responses.
Chu S, et al.
International Immunopharmacology, 19(2), 317-326 (2014)
Inhibitory effect of ginsenoside Rg5 and its metabolite ginsenoside Rh3 in an oxazolone-induced mouse chronic dermatitis model.
Shin W-Y, et al.
Archives of Pharmacal Research, 29(8), 685-690 (2006)
Simultaneous quantification of 14 ginsenosides in Panax ginseng CA Meyer (Korean red ginseng) by HPLC-ELSD and its application to quality control.
Kim NS, et al.
Journal of Pharmaceutical and Biomedical Analysis, 45(1), 164-170 (2007)

Articles

In this article we present several HPTLC applications and analytical standards for ginsenosides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service