43052
Müller-Kauffmann Tetrathionate Broth, Base (ISO)
suitable for microbiology, NutriSelect® Plus
Synonym(s):
Tetrathionate Müller-Kaufmann Broth Base
About This Item
Recommended Products
Agency
according to ISO 19250:2010
according to ISO 6579-1:2017
Quality Level
sterility
non-sterile
form
powder
shelf life
limited shelf life, expiry date on the label
composition
bile salts, 4.78 g/L
calcium carbonate, 38.7 g/L
casein peptone, 8.60 g/L
meat extract, 4.30 g/L
sodium chloride, 2.60 g/L
sodium thiosulfate, 47.80 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
application(s)
clinical testing
environmental
food and beverages
microbiology
suitability
selective for Salmonella spp.
selective for Shigella spp.
General description
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service