Skip to Content
Merck
All Photos(1)

Key Documents

42488

Supelco

Bacopaside X

analytical standard

Synonym(s):

Jujubogenin 3-O-[β-D-glucopyranosyl-(1→3)-[α-L-arabinofuranosyl-(1→2)]-α-L-arabinopyranoside], Jujubogenin isomer of bacopasaponin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H74O17
CAS Number:
Molecular Weight:
899.07
UNSPSC Code:
85151701
Pricing and availability is not currently available.

grade

analytical standard

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

InChI

1S/C46H74O17/c1-21(2)14-22-15-44(7,55)37-23-8-9-28-42(5)12-11-29(41(3,4)27(42)10-13-43(28,6)45(23)19-46(37,63-22)57-20-45)60-40-36(62-38-33(53)31(51)26(17-48)59-38)35(24(49)18-56-40)61-39-34(54)32(52)30(50)25(16-47)58-39/h14,22-40,47-55H,8-13,15-20H2,1-7H3/t22-,23+,24-,25+,26-,27-,28+,29-,30+,31-,32-,33+,34+,35-,36+,37-,38-,39-,40-,42-,43+,44-,45-,46-/m0/s1

InChI key

RANQPHKSRUUPKK-GPUGMLHBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitra Nuengchamnong et al.
Journal of pharmaceutical and biomedical analysis, 129, 121-134 (2016-07-17)
Bacopa monnieri (L.) Wettst. (Scrophulariaceae) is an Ayurvedic medicinal plant used as a memory enhancer. Its major chemical constituents are Bacopa saponins consisting of jujubogenin and pseudojujubogenin glycosides. These two aglycones are isomers different at the positions of prenyl substitution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service