Skip to Content
Merck
All Photos(1)

Key Documents

42077

SAFC

Sodium cyanoborohydride

Pharma Manufacturing

Synonym(s):

Sodium cyanotrihydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH3CN
CAS Number:
Molecular Weight:
62.84
Beilstein:
4152656
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥97% (wt., redox titration)

form

powder

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: reductant

impurities

≤1000 ppm residual solvent (tetrahydrofuran)
≤3000 ppm residual solvent (toluene)

mp

>242 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL, clear to faintly turbid, colorless

anion traces

(may contain traces of cyanide and borohydride)

cation traces

Ag: ≤2.5 mg/kg
Cd: ≤2.5 mg/kg
Cr: ≤2.5 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Mo: ≤2.5 mg/L
Ni: ≤2.5 mg/kg

suitability

suitable for manufacturing use

SMILES string

[Na+].[BH3-]C#N

InChI

1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1

InChI key

CVDUGUOQTVTBJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koen van der Maaden et al.
Langmuir : the ACS journal of surfaces and colloids, 31(31), 8654-8660 (2015-07-07)
The aim of this work was to coat pH-sensitive microneedle arrays with inactivated polio vaccine (IPV) particles and N-trimethyl chitosan chloride (TMC) via electrostatic interactions, and assess the immunogenicity of the vaccine after topical application of the coated microneedles in
Lowell J Brady et al.
mAbs, 7(3), 562-570 (2015-04-22)
Accurate measurement and functional characterization of antibody Fc domain N-linked glycans is critical to successful biosimilar development. Here, we describe the application of methods to accurately quantify and characterize the N-linked glycans of 2 IgG1 biosimilars with effector function activity
Hwiwon Lee et al.
ACS nano, 8(5), 4790-4798 (2014-04-16)
Rheumatoid arthritis (RA) is a chronic inflammatory immune disease causing the inflammation of synovial membrane and the articular cartilage destruction. In this work, hyaluronate-gold nanoparticle/Tocilizumab (HA-AuNP/TCZ) complex was prepared for the treatment of RA. AuNP was used as a drug
Ruby Bansal et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 87(3), 461-471 (2014-05-16)
Intracellular nature and diversified locations of infectious and parasitic diseases such as leishmaniasis, trypanosomiasis, tuberculosis and hepatitis B and C pose a significant global burden and challenge to the scientists working in the area of drug discovery and drug delivery.
Jana Kucerova et al.
Materials science & engineering. C, Materials for biological applications, 40, 308-315 (2014-05-27)
In this study, magnetic poly(glycidyl methacrylate) microparticles containing carboxyl groups (PGMA-COOH) were coated using highly hydrophilic polymer poly(ethylene glycol) (PEG). PEG was used to reduce nonspecific interactions with proteins and cells while decreasing adhesion of particles to the walls of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service