Skip to Content
Merck
All Photos(1)

Key Documents

38719

Millipore

Photobacterium Broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€354.00

€354.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€354.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

€354.00


Please contact Customer Service for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

ammonium chloride, 0.3 g/L
calcium carbonate, 1 g/L
casein enzymic hydrolisate, 5 g/L
ferric chloride, 0.01 g/L
magnesium sulfate, 0.3 g/L
monopotassium dihydrogen phosphate, 3 g/L
sodium chloride, 30 g/L
sodium glycerophosphate, 23.5 g/L
yeast extract, 2.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
life science and biopharma

microbiology

suitability

photobacteria

Application

Photobacterium Broth has been used to enumerate Photobacterium sp..[1][2]
Photobacterium Broth is used for cultivation and demonstration of luminescence of photobacteria.

Preparation Note

Suspend 65.6 g in 1 litre distilled water. Heat to boiling to dissolve the medium. Sterilize by autoclaving at 121°C for 15 minutes. Before pouring mix well to evenly distribute slight precipitate formed.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin Fogarty et al.
Food microbiology, 77, 38-42 (2018-10-10)
This study investigated the growth of indicator and spoilage bacteria on whole Atlantic salmon (Salmo salar) stored aerobically at 2 °C. On days 0, 2, 3, 6, 8 and 10 microbiological analysis was carried out on inner flesh and outer skin
The Effect of Organic Acid, Trisodium Phosphate and Essential Oil Component Immersion Treatments on the Microbiology of Cod (Gadus morhua) during Chilled Storage.
Smyth, et al.
Foods (Basel, Switzerland), 7 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service