Skip to Content
Merck
All Photos(2)

Key Documents

37087

Supelco

Tetraconazole

PESTANAL®, analytical standard

Synonym(s):

(RS)-2-(2,4-Dichlorophenyl)-3-(1H-1,2,4-triazolyl)propyl 1,1,2,2-tetrafluoroethyl ether, (±)-1-[2-(2,4-Dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-1H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€232.00

€232.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€232.00

About This Item

Empirical Formula (Hill Notation):
C13H11Cl2F4N3O
CAS Number:
Molecular Weight:
372.15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€232.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

FC(F)C(F)(F)OCC(Cn1cncn1)c2ccc(Cl)cc2Cl

InChI

1S/C13H11Cl2F4N3O/c14-9-1-2-10(11(15)3-9)8(4-22-7-20-6-21-22)5-23-13(18,19)12(16)17/h1-3,6-8,12H,4-5H2

InChI key

LQDARGUHUSPFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetraconazole is a systemic[1]broad spectrum triazole fungicide[1], which can be used to control a large number of fungi like Ascomycetes, Basidiomycetes, and Deuteromycetes especially Sphaerotheca fuliginea on cucumber and Erysiphe graminis on wheat.[2] It can be prepared, via stereoselective lipase-catalyzed hydrolysis of the racemic precursor (R,S)-2-(2,4-dichlorophenyl)-3-(1H-1,2,4-triazol-l-y1)-prop-l-yl acetate.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemoenzymic synthesis and biological activity of both enantiomeric forms of tetraconazole, a new antifungal triazole
Bianchi D, et al.
Journal of Agricultural and Food Chemistry, 39, 197-201 (1991)
Ultrastructural studies of the effects of tetraconazole on the barley-powdery mildew host pathogen complex
Maffi.D, et al.
Mycological research, 99, 799-805 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service