Skip to Content
Merck
All Photos(2)

Key Documents

37046

Supelco

Butamifos

PESTANAL®, analytical standard

Synonym(s):

O-Ethyl O-(5-methyl-2-nitrophenyl) N-sec-butylphosphorothionoamidate, O-Ethyl O-6-nitro-m-tolyl sec-butylphosphoramidothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21N2O4PS
CAS Number:
Molecular Weight:
332.36
Beilstein:
2165895
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(NC(C)CC)Oc1cc(C)ccc1[N+]([O-])=O

InChI

1S/C13H21N2O4PS/c1-5-11(4)14-20(21,18-6-2)19-13-9-10(3)7-8-12(13)15(16)17/h7-9,11H,5-6H2,1-4H3,(H,14,21)

InChI key

OEYOMNZEMCPTKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butamifos belongs to the class of phosphoramidothioate herbicides used in both crop and non-crop areas.

Application

Butamifos may be used as an analytical reference standard for the quantification of the analyte in environmental and tea samples using various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic Pathways of Agrochemicals: Herbicides and Plant Growth Regulators (2007)
Simultaneous determination of 103 pesticide residues in tea samples by LC-MS/MS.
Huang z, et al.
Journal of Separation Science, 32(9), 1294-1301 (2009)
Simple GC and GC/MS determination of Herbicides in air using quartz fiber filter and activated carbon fiber filter for sampling
Kawata K
Journal of Environmental Chemistry, 2(2), 181-186 (1992)
Laura Morello et al.
Antimicrobial agents and chemotherapy, 64(3) (2019-12-25)
Microorganisms belonging to the genus Prototheca are achlorophyllous microalgae, occasionally behaving as environmental pathogens that cause severe mastitis in milk cows, as well as localized or systemic infections in humans and animals. Among the different species belonging to the genus
Simultaneous determination of 64 pesticides in river water by stir bar sorptive extraction and thermal desorption-gas chromatography-mass spectrometry
Nakamura S and Daishima S
Analytical and Bioanalytical Chemistry, 382(1), 99-107 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service