Skip to Content
Merck
All Photos(1)

Key Documents

37021

Supelco

L-Hyoscyamine

analytical standard

Synonym(s):

[3(S)-endo]-α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO3
CAS Number:
Molecular Weight:
289.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CN1C2CCC1CC(C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3

InChI key

RKUNBYITZUJHSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

For the analysis of the phytotoxins. Tropane alkaloid.

Biochem/physiol Actions

Competitive antagonist at post-ganglionic synapses and on smooth muscle

Reconstitution

Dried down, ~100 μg/mL after reconstitution with 1 mL of water

Analysis Note

purity : ≥96.0% (HPLC)

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Wen Cheng et al.
International journal of medicinal mushrooms, 15(3), 277-286 (2013-05-15)
In our previous research, Cordyceps militaris (CM) had a hypoglycemic effect in normal rats. In this study we wanted to elucidate whether CM also had an effect on diabetic rats. Twelve rats with streptozotocin-induced diabetes were separated randomly into 2
Kammi B Gunton
Pediatrics, 131(3), 540-547 (2013-02-06)
Amblyopia is the most common cause of preventable visual loss in children. This article reviews treatment options, durations, and efficacy in randomized multicentered trials conducted by the Pediatric Eye Disease and Investigator Group in the last decade. Parents and patients
Marija Lukac et al.
European journal of pediatric surgery : official journal of Austrian Association of Pediatric Surgery ... [et al] = Zeitschrift fur Kinderchirurgie, 23(1), 80-84 (2013-02-05)
Evaluation of the effectiveness of oral atropine versus surgical therapy for hypertrophic pyloric stenosis (HPS). A total of 66 consecutive patients with HPS were treated at the University Children's Hospital between January 2006 and December 2011. The diagnosis was initially
Peter Jones et al.
PloS one, 8(2), e57478-e57478 (2013-03-08)
Atropine has is currently recommended to facilitate haemodynamic stability during critical care intubation. Our objective was to determine whether atropine use at induction influences ICU mortality. A 2-year prospective, observational study of all first non-planned intubations, September 2007-9 in PICU
Max J Hilz et al.
Stroke, 44(4), 1062-1069 (2013-02-21)
Pharmacological blockade showed sympathetic origin of 0.03 to 0.15 Hz blood pressure (BP) oscillations and parasympathetic origin of 0.15 to 0.5 Hz RR-interval (RRI) oscillations, but has not been used to determine origin of cerebral blood flow velocity (CBFV) oscillations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service