Skip to Content
Merck
All Photos(1)

Key Documents

36841

Supelco

Diisodecyl adipate

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H21O2C(CH2)4CO2C10H21
CAS Number:
Molecular Weight:
426.67
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Selectophore

Quality Level

form

liquid

refractive index

n20/D 1.452 (lit.)
n20/D 1.452

mp

27.4 °C (lit.)

density

0.916 g/mL at 20 °C
0.916 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCCCOC(=O)CCCCC(=O)OCCCCCCCC(C)C

InChI

1S/C26H50O4/c1-23(2)17-11-7-5-9-15-21-29-25(27)19-13-14-20-26(28)30-22-16-10-6-8-12-18-24(3)4/h23-24H,5-22H2,1-4H3

InChI key

YKGYQYOQRGPFTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

429.8 °F - closed cup

Flash Point(C)

221 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard F Grossman
Handbook of Vinyl Formulating, 192-192 (2008)
Sol-Gel Method for the Matrix of Chloride-Selective Membranes Doped with Tridodecylmethylammonium Chloride
Kim, W., et al.
Analytical Chemistry, 69, 95-96 (1997)
T Nørbygaard et al.
Applied spectroscopy, 58(4), 410-413 (2004-04-24)
Fourier transform (FT) Raman spectroscopy is applied to a range of phthalate ester plasticizers in pure form as well as in poly(vinyl chloride) (PVC) samples. It is found that phthalate esters as a group can be identified by a set

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service