Skip to Content
Merck
All Photos(1)

Key Documents

36788

Sigma-Aldrich

4-(2,5-Diethoxy-4-nitrophenyl)morpholine

purum, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O5
CAS Number:
Molecular Weight:
296.32
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥99.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

very deep yellow

mp

137-141 °C (lit.)
137-141 °C

SMILES string

CCOc1cc(c(OCC)cc1N2CCOCC2)[N+]([O-])=O

InChI

1S/C14H20N2O5/c1-3-20-13-10-12(16(17)18)14(21-4-2)9-11(13)15-5-7-19-8-6-15/h9-10H,3-8H2,1-2H3

InChI key

KNHGNICBXADRMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service