Skip to Content
Merck
All Photos(1)

Documents

36545

Supelco

Dichloroacetic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCOOH
CAS Number:
Molecular Weight:
128.94
Beilstein:
1098596
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

4.5 (vs air)

vapor pressure

0.19 mmHg ( 20 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.466 (lit.)

bp

194 °C (lit.)

mp

9-11 °C (lit.)

density

1.563 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)C(Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

JXTHNDFMNIQAHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Brain,Liver,Testes

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haihua Xiao et al.
Chemical communications (Cambridge, England), 48(87), 10730-10732 (2012-09-27)
A multifunctional hybrid platinum(IV) prodrug, which consists of both the mitochondria-targeting drug DCA and the DNA-crosslinking drug cisplatin, was synthesized and tethered to a carrier polymer to further self-assemble into micelles for intracellular delivery.
Yong Won Choi et al.
Cancer letters, 346(2), 300-308 (2014-02-01)
To investigate sensitization of metformin-cytotoxicity, cancer cells were treated with dichloroacetate (DCA), an inhibitor of pyruvate dehydrogenase kinase (PDK). Metformin-cytotoxicity was mainly dependent on glucose availability and reducing power generated by pentose phosphate pathway, whereas DCA cotreatment enhanced metformin-cytotoxicity via
Toshimitsu Ohashi et al.
International journal of cancer, 133(5), 1107-1118 (2013-02-20)
The activation of oncogenic signaling pathways induces the reprogramming of glucose metabolism in tumor cells and increases lactic acid secretion into the tumor microenvironment. This is a well-known characteristic of tumor cells, termed the Warburg effect, and is a candidate
Yi-Hsueh Chuang et al.
Water research, 47(3), 1308-1316 (2013-01-05)
The direct incorporation of chloramines and dissolved organic nitrogen (DON) may provide the nitrogen for nitrogenous disinfection byproducts (N-DBPs). This study explores the contributions of natural DON and chloramine incorporation to the formation of N-DBPs during chloramination. This study also
Mehdi D Esrafili
Journal of molecular modeling, 18(12), 5005-5016 (2012-06-28)
A theoretical study was performed to examine hydrogen and halogen bonds properties in gas phase and crystalline dichloroacetic acid (DCAA). The specific pattern of O-H∙∙∙O, C-H∙∙∙O, HCl, Cl∙∙∙O and Cl∙∙∙Cl interactions in DCAA dimers is described within the quantum theory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service