Skip to Content
Merck
All Photos(1)

Documents

36521

Supelco

Quinchlorac

PESTANAL®, analytical standard

Synonym(s):

3,7-Dichloro-8-quinolinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Cl2NO2
CAS Number:
Molecular Weight:
242.06
Beilstein:
7761858
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1c(Cl)ccc2cc(Cl)cnc12

InChI

1S/C10H5Cl2NO2/c11-6-3-5-1-2-7(12)8(10(14)15)9(5)13-4-6/h1-4H,(H,14,15)

InChI key

FFSSWMQPCJRCRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinchlorac is a postemergence herbicide, used for the control of broadleaf weeds, especially white clover (Trifolium repens L.) and Veronica filiformis.

Application

Quinchlorac may be used as an analytical reference standard for the quantification of the analyte in environmental samples using high-performance liquid chromatography with ultraviolet detection (HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen-Mei Lu et al.
Biomedical and environmental sciences : BES, 16(4), 314-322 (2004-03-12)
To investigate the potential effects of herbicide quinclorac (3,7-dichloro-8-quinoline-carboxylic) on the culturable microorganisms in flooded paddy soil. Total soil aerobic bacteria, actinomycetes and fungi were counted by a 10-fold serial dilution plate technique. Numbers of anaerobic fermentative bacteria (AFB), denitrifying
Zhenmei Lü et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 38(6), 771-782 (2003-12-03)
Strain WZI capable of degrading quinclorac was isolated from a pesticide manufactory soil and considered to be Burkholderia cepacia, belonged to bacteria, Proteobacteria, beta-Proteobacteria, based on morphology, physio-biochemical properties, whole cell fatty acid analysis and a partial sequencing of 16S
Development and validation of a high-performance liquid chromatographic procedure for the determination of herbicide residues in surface and agriculture waters
Zanella R, et al.
Journal of Separation Science, 26(9-10), 935-938 (2003)
N Mito et al.
Plant physiology, 109(1), 293-297 (1995-09-01)
The effect of a tomato (Lycopersicon esculentum) mutation, diageotropica (dgt), on the accumulation of mRNA corresponding to tomato homologs of three auxin-regulated genes, LeAux, LeSAUR, and Lepar, was examined. The dgt mutation inhibited the induction of LeAux and LeSAUR mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service