Skip to Content
Merck
All Photos(1)

Key Documents

36333

Supelco

3-Hydroxybenzoic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
508160
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

200-203 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(=O)c1cccc(O)c1

InChI

1S/C7H6O3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

IJFXRHURBJZNAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kayleigh A Clarke et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 972, 29-37 (2014-10-13)
The simultaneous analysis of free-form and conjugated flavonoids in the same sample is difficult but necessary to properly estimate their bioavailability. A method was developed to optimise the extraction of both free and conjugated forms of catechins and metabolites in
Pradeepraj Durairaj et al.
Enzyme and microbial technology, 70, 58-65 (2015-02-11)
FoCYP53A19, a novel cytochrome P450 capable of performing benzoate hydroxylation, was identified and characterized from the ascomycete Fusarium oxysporum f.sp. lycopersici. Comparative functional analysis of FoCYP53A19 with the heterologous and homologous cytochrome P450 reductases (CPR) such as Saccharomyces cerevisiae (ScCPR)
Katheryn M Goodrich et al.
Journal of agricultural and food chemistry, 62(46), 11190-11199 (2014-10-23)
Metabolism of flavanols (catechins, procyanidins) by gut microbiota has been extensively characterized. Comparatively little is known about accumulation of flavanols and their metabolites in the colon tissues, particularly during chronic exposure to low doses. Mice were fed low doses of
Pankaj Aggarwal et al.
Journal of chromatography. A, 1364, 96-106 (2014-09-07)
Highly cross-linked monolithic networks (i.e., polyethylene glycol diacrylate, PEGDA) synthesized from monomers containing varying ethylene oxide chain lengths were fabricated inside fused silica capillary columns for use in liquid chromatography (LC) of small molecules. Tergitol was used as a surfactant
Mingqi Xie et al.
Nucleic acids research, 42(14), e116-e116 (2014-07-18)
Synthetic biology has significantly advanced the design of mammalian trigger-inducible transgene-control devices that are able to programme complex cellular behaviour. Fruit-based benzoate derivatives licensed as food additives, such as flavours (e.g. vanillate) and preservatives (e.g. benzoate), are a particularly attractive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service