Skip to Content
Merck
All Photos(1)

Documents

36131

Supelco

Ioxynil

PESTANAL®, analytical standard

Synonym(s):

4-Hydroxy-3,5-diiodobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3I2NO
CAS Number:
Molecular Weight:
370.91
Beilstein:
2364041
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Oc1c(I)cc(cc1I)C#N

InChI

1S/C7H3I2NO/c8-5-1-4(3-10)2-6(9)7(5)11/h1-2,11H

InChI key

NRXQIUSYPAHGNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ioxynil is a post-emergent herbicide, which can be used to selectively control broad-leaved weeds in cereals and grass crops. It can be used in combination with phenoxy herbicides in order to control hormone-resistant weeds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modern Weed Control (1975)
Carsten T Petersen et al.
Pest management science, 59(1), 85-96 (2003-02-01)
Knowledge of the movement of herbicides and soil particles to sub-surface tile drainage may help to predict chemical leaching to surface waters and deeper groundwater systems. The movement of pendimethalin (2 years), ioxynil (1 year) and soil particles (3 years)
Edward Leithe et al.
Toxicology and applied pharmacology, 247(1), 10-17 (2010-06-01)
Gap junctions are intercellular plasma membrane domains containing channels that mediate transport of ions, metabolites and small signaling molecules between adjacent cells. Gap junctions play important roles in a variety of cellular processes, including regulation of cell growth and differentiation
M Millet et al.
Ecotoxicology and environmental safety, 41(1), 44-50 (1998-10-03)
This paper presents first experiments of laboratory investigations of the photodegradation by direct photolysis (lambda>290 nm) of cholorotahlonil, dichlobenil, chloroxynil, bromoxynil, and ioxynil in aqueous, pH-buffered, and organic solutions and the calculation of the quantum yields. The photolysis of chlorothalonil
Pesticide Chemistry (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service