Skip to Content
Merck
All Photos(2)

Key Documents

35835

Supelco

2,5-Dichlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
1907692
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

211 °C (lit.)

mp

54-57 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1cc(Cl)ccc1Cl

InChI

1S/C6H4Cl2O/c7-4-1-2-5(8)6(9)3-4/h1-3,9H

InChI key

RANCECPPZPIPNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Dichlorophenol may be used as an analytical reference standard for the determination of the analyte in water,[1] and wood, paper, cardboard, fruits, and fruit juices[2] using simultaneous dispersive liquid-liquid microextraction and derivatization followed by gas chromatography-electron-capture detection (GC-ECD) and gas chromatography-mass spectrometry (GC-MS), respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R H Hill et al.
Archives of environmental health, 50(4), 277-280 (1995-07-01)
p-Dichlorobenzene is used widely in the United States as a room deodorizer, a moth repellent, and a precursor for a polymer. In a previous study of selected children in Arkansas, we found that 96% of the children had detectable urinary
Xiaoyun Ye et al.
Environment international, 35(8), 1160-1163 (2009-08-12)
In humans, the metabolism of environmental phenols may include the formation of conjugated species (e.g., glucuronides and sulfates), but the free species-not the conjugated forms-are considered biologically active. Therefore, information on the concentration of these free species in blood or
M A Sogorb et al.
Chemico-biological interactions, 87(1-3), 117-125 (1993-06-01)
The organophosphorus (OP) compound O-hexyl-O-2,5-dichlorophenyl phosphoramidate (H-DCP) is hydrolysed in the plasma, liver and brain of hens and rats. We study in hen plasma the effect of tissue and substrate concentrations and of pH on the hydrolysing activity of H-DCP.
N Díaz-Alejo et al.
Journal of chromatography, 622(2), 179-186 (1993-12-22)
O-Hexyl O-2,5-dichlorophenyl phosphoramidate (HDCP) is a chiral organophosphorus compound that undergoes enzymatic hydrolysis in the rat and hen. Studies of the stereospecificity of its biodegradation are necessary to establish HDCP toxicity. To this effect, methods have been developed for the
J S Holler et al.
Journal of analytical toxicology, 13(3), 152-157 (1989-05-01)
We have developed a method for determining selected chlorinated phenols and phenoxy herbicides in urine. The process of preparing the samples includes acid hydrolysis, extraction with benzene, derivatization with diazoethane, and column chromatography cleanup. We quantify the more volatile compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service