Skip to Content
Merck
All Photos(2)

Documents

35378

Supelco

Haloxyfop-P

PESTANAL®, analytical standard

Synonym(s):

(R)-2-[4-(3-Chloro-5-(trifluoromethyl)-2-pyridyloxy)phenoxy]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70
Beilstein:
8571970
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C[C@@H](Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1)C(O)=O

InChI

1S/C15H11ClF3NO4/c1-8(14(21)22)23-10-2-4-11(5-3-10)24-13-12(16)6-9(7-20-13)15(17,18)19/h2-8H,1H3,(H,21,22)/t8-/m1/s1

InChI key

GOCUAJYOYBLQRH-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Haloxyfop-P is a postemergence herbicide, primarily used for the control of grass weeds in rice cultivation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Xu et al.
Journal of fluorescence, 27(6), 2195-2200 (2017-09-13)
The wide use of pesticide p-fluorophenoxyacetic acid has caused the serious environmental contaminant. A novel fluorescent probe for sensitive detection of p-fluorophenoxyacetic acid in aqueous solutions based on 3.0G quaternary ammonium polyamidoamine (PAMAM) dendrimer modified quantum dots (QDs) (PAMAM@QDs) was
Efficacy and crop safety of several cotton herbicides [J].
Lian Y-Z, et al.
Agrochemicals, 4, 17-17 (2006)
Yefei Xu et al.
Ecotoxicology and environmental safety, 189, 110003-110003 (2019-12-04)
When the amount of pesticide exceeds the self-purification ability of the environment, it will be enriched in the human body through the atmosphere, soil, water circulation, etc., threatening human health. Aryloxy-phenoxy-propionate (APP) herbicides are a class of acetyl-CoA carboxylase (ACCase)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service