Skip to Content
Merck
All Photos(1)

Key Documents

34502

Supelco

Prometryn-(N2-isopropyl-d7)

PESTANAL®, analytical standard

Synonym(s):

2-(Isopropyl-d7-amino)-4-(isopropylamino)-6-methylthio-1,3,5-triazine, N2-(1-Methylethyl-d7)-N4-(1-methylethyl)-6-(methylthio)-1,3,5-triazine-2,4-diamine, Prometryn-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D7H12N5S
CAS Number:
Molecular Weight:
248.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(C)NC1=NC(SC)=NC(NC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=N1

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Betty Chaumet et al.
Ecotoxicology (London, England), 29(2), 185-195 (2020-01-24)
Variations of temperature and photoperiod throughout different seasons can affect aquatic communities such as biofilms. Biofilms, generally present at the base of trophic chains in freshwaters, are also subject to organic contamination, and are especially affected by herbicides. Many studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service