Skip to Content
Merck
All Photos(1)

Key Documents

34341

Supelco

Famphur

PESTANAL®, analytical standard

Synonym(s):

O-(4-Dimethylsulfamoylphenyl)-O,O-dimethyl-phosphorothioate, Famophos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16NO5PS2
CAS Number:
Molecular Weight:
325.34
Beilstein:
2224254
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COP(=S)(OC)Oc1ccc(cc1)S(=O)(=O)N(C)C

InChI

1S/C10H16NO5PS2/c1-11(2)19(12,13)10-7-5-9(6-8-10)16-17(18,14-3)15-4/h5-8H,1-4H3

InChI key

JISACBWYRJHSMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Famphur is an organophosphate insecticide used primarily as a veterinary drug for the control of warbles in cattle.[1]

Application

Famphur may be used as an analytical reference standard for the determination of the analyte in animal muscles[2] and rice samples[3] using liquid chromatography coupled with electrospray ionization tandem mass spectrometry (LC/MS/MS), and QuEChERS extraction and gas chromatography-tandem mass Spectrometry, respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F - closed cup

Flash Point(C)

> 100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organophosphate insecticide (famphur) topically applied to cattle kills magpies and hawks.
Henny CJ, et al.
The Journal of wildlife management, 648-658 (1985)
J I Cacho et al.
Journal of chromatography. A, 1559, 95-101 (2018-01-02)
Nine organophosphorus pesticides (OPPs) were determined in environmental waters from different origins using in situ ionic liquid dispersive liquid microextraction (IL-DLLME). This preconcentration technique was coupled to gas chromatography-mass spectrometry (GC-MS) using microvial insert thermal desorption, an approach that uses
A multi-residue method for the determination of 124 pesticides in rice by modified QuEChERS extraction and gas chromatography-tandem mass spectrometry.
Hou X, et al.
Food Chemistry, 138(2-3), 1198-1205 (2013)
C J Henny et al.
Journal of wildlife diseases, 23(2), 292-295 (1987-04-01)
Since 1982 when secondary poisoning of a red-tailed hawk (Buteo jamaicensis) was documented following the recommended use of famphur applied topically to cattle, the Patuxent Wildlife Research Center has tested dead birds of prey for poisoning by famphur and other
P Eyre et al.
American journal of veterinary research, 42(1), 25-28 (1981-01-01)
Soluble toxin of Hypoderma lineatum injected IV induced a severe cardiorespiratory shock syndrome in calves. Intradermal injection of the toxin caused well-defined, edematous wheals comparable with those produced by histamine. Treatment of calves with famphur (organophosphate insecticide; poured on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service