Skip to Content
Merck
All Photos(1)

Key Documents

34033

Supelco

Aflatoxin G2 solution

0.5 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€295.00

€295.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
€295.00

About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Beilstein:
1692017
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€295.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

0.5 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

COc1cc2O[C@H]3OCC[C@H]3c2c4OC(=O)C5=C(CCOC5=O)c14

InChI

1S/C17H14O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WPCVRWVBBXIRMA-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aflatoxins are fungal secondary toxic metabolites produced by Aspergillus flavus and Aspergillus parasiticus, which display mutagenicity, teratogenicity and carcinogenicity.[1][2]

Application

Aflatoxin G2 may be used as a reference standard in the determination of the analyte:
  • In medicinal herbs by liquid chromatography coupled with tandem mass spectrometry.[2]
  • In animal feed by ultra-high performance liquid chromatography coupled with tandem mass spectrometry.[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of aflatoxins B1, G1, B2 and G2 in medicinal herbs by liquid chromatography- tandem mass spectrometry.
Ventura M, et al.
Journal of Chromatography A, 1048(1), 25-29 (2004)
Inhibition of aflatoxin B1, B2, G1 and G2 production by Aspergillus parasiticus in nuts using yellow and oriental mustard flours.
Hontanaya C, et al.
Food Control, 47, 154-160 (2015)
Robert A Everley et al.
Journal of analytical toxicology, 31(3), 150-156 (2007-06-21)
Automated immunoaffinity solid-phase extraction followed by liquid chromatography-tandem mass spectrometry and chemical analogue internal standardization is employed to detect and quantify the aflatoxins AFB(1), AFB(2), AFG(1), AFG(2), and the metabolites AFM(1) and AFP(1) in urine. The dynamic range of the
Viresh Mohanlall et al.
Journal of food protection, 69(9), 2224-2229 (2006-09-26)
Phytoalexins (stress-induced compounds) from Citrus sinensis cultivar Valencia were screened for antifungal and antimycotoxic activity against a test organism (Cladosporium cladosporoides) and mycotoxin-producing fungi Fusarium verticillioides and Aspergillus parasiticus. The active compound, a member of the coumarin family of compounds
J Tam et al.
Food additives and contaminants, 23(7), 693-699 (2006-06-06)
Three hundred and forty-nine breakfast and infant cereal samples were collected at retail level across Canada from 2002 to 2005. They included rice-, soy-, barley-based and mixed-grain infant cereals, corn-, wheat-, rice-based and mixed-grain breakfast cereals, and were analysed for

Protocols

LC/MS/MS Analysis of Aflatoxins in Hops on Ascentis® Express 2.7 μm Phenyl-Hexyl after Cleanup Using Supel™ Tox AflaZea SPE

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service