Skip to Content
Merck
All Photos(1)

Key Documents

33997

Supelco

Imazalil sulfate

PESTANAL®, analytical standard

Synonym(s):

1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy)ethyl]-1H-imidazole sulfate, Enilconazole sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14Cl2N2O · H2SO4
CAS Number:
Molecular Weight:
395.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OS(O)(=O)=O.Clc1ccc(C(Cn2ccnc2)OCC=C)c(Cl)c1

InChI

1S/C14H14Cl2N2O.H2O4S/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16;1-5(2,3)4/h2-6,8,10,14H,1,7,9H2;(H2,1,2,3,4)

InChI key

XVTXMTOYQVRHSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imazalil sulfate is an imidazole fungicide and its mode of action involves the inhibition of sterol synthesis thus affecting cell wall permeability in fungi.[1]

Application

Imazalil sulfate may be used as a standard test material to investigate its electrochemical degradation by anodic oxidation on boron-doped diamond (BDD) electrode.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical degradation of imazalil and pyrimethanil by anodic oxidation on boron-doped diamond.
Errami M, et al.
Journal of Chemical and Pharmaceutical Research, 4(7), 3518-3518 (2012)
E J Keeble et al.
The Veterinary record, 167(22), 862-864 (2011-01-26)
An outbreak of Trichophyton dermatophytosis was diagnosed in a group of four L'Hoest's monkeys (Cercopithecus lhoesti) housed in the primate section at a zoological collection. The affected animals presented with areas of non-pruritic alopecia, scaling and crusting. The diagnosis was
Hasan Türkez et al.
Toxicology and industrial health, 28(7), 648-654 (2011-10-12)
Ascorbic acid (AA), known as vitamin C, has important antioxidant and metabolic functions, making its incorporation into the human diet essential. On the other hand, imazalil (IMA), a commonly used fungicide in both agricultural and clinical domains is suspected to
[Malignant melanophoroma in a Hermann's tortoise (Testudo hermanni). A case report].
K O Heckers et al.
Tierarztliche Praxis. Ausgabe K, Kleintiere/Heimtiere, 39(1), 45-50 (2011-12-07)
M Sharman et al.
The Journal of small animal practice, 53(3), 161-167 (2012-02-04)
Multiple topical treatments are often required for clinical cure of mycotic rhinosinusitis in dogs. The objective of this study was to describe the distribution and retention of enilconazole and clotrimazole solutions using a temporary trephination protocol. Nine client-owned dogs diagnosed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service