Skip to Content
Merck
All Photos(1)

Key Documents

33910

Sigma-Aldrich

1,2-Dibromoethylene

purum, cis+trans, ≥98.0% (GC)

Synonym(s):

Acetylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH=CHBr
CAS Number:
Molecular Weight:
185.85
Beilstein:
1719351
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.542

bp

107-110 °C (lit.)

density

2.27 g/mL at 20 °C (lit.)

SMILES string

Br\C=C\Br

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service