Skip to Content
Merck
All Photos(1)

Key Documents

33567

Supelco

4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 monoethoxylate solution

10 μg/mL in acetone, analytical standard

Synonym(s):

2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy-3,5-d2]ethanol, 2-[4-(3,6-Dimethyl-3-heptyl)phenoxy-3,5-d2]ethanol, 3,6,3-Nonylphenol-monoethoxylate, 363-NP1EO-D2, 4-(1-Ethyl-1,4-dimethylpentyl)phenol-3,5-d2-monoethoxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D2H26O2
CAS Number:
Molecular Weight:
266.42
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+2

storage temp.

−20°C

SMILES string

[2H]c1cc(OCCO)cc([2H])c1C(C)(CC)CCC(C)C

InChI

1S/C17H28O2/c1-5-17(4,11-10-14(2)3)15-6-8-16(9-7-15)19-13-12-18/h6-9,14,18H,5,10-13H2,1-4H3/i6D,7D

InChI key

QRFCMJANHDRHEH-QFIQSOQBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - Information refers to the main ingredient.

Flash Point(C)

-17 °C - Information refers to the main ingredient.


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XIV (Authorisation List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anika L Dzierlenga et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(6), 722-732 (2019-11-05)
Poly- and perfluorinated alkyl substances (PFAS) are environmentally persistent chemicals associated with many adverse health outcomes. The National Toxicology Program evaluated the toxicokinetics (TK) of several PFAS to provide context for toxicologic findings.Plasma TK parameters and tissue (liver, kidney, brain)

Protocols

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service