Skip to Content
Merck
All Photos(1)

Key Documents

33365

Supelco

Hexythiazox

PESTANAL®, analytical standard

Synonym(s):

trans-5-(4-Chlorophenyl)-N-cyclohexyl-4-methyl-2-oxothiazolidine-3-carboxamide)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€78.70

€78.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€78.70

About This Item

Empirical Formula (Hill Notation):
C17H21ClN2O2S
CAS Number:
Molecular Weight:
352.88
Beilstein:
9001643
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€78.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

106-109 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

C[C@H]1[C@@H](SC(=O)N1C(=O)NC2CCCCC2)c3ccc(Cl)cc3

InChI

1S/C17H21ClN2O2S/c1-11-15(12-7-9-13(18)10-8-12)23-17(22)20(11)16(21)19-14-5-3-2-4-6-14/h7-11,14-15H,2-6H2,1H3,(H,19,21)/t11-,15+/m0/s1

InChI key

XGWIJUOSCAQSSV-XHDPSFHLSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hexythiazox is an acaricide used for controlling mite growth on major crops such as pear, apple, citrus, tree nuts, stone fruits, cranberries, non-bearing trees, vines, strawberries, cotton, hop, mint, ornamental landscape plantings, orchids, and alfalfa.[1]

Application

Hexythiazox may be used as a reference standard for the determination of hexythiazox in fruits and vegetables by supercritical fluid extraction and high-performance liquid chromatography (HPLC) method.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive assay of hexythiazox residue in citrus fruits using gold nanoparticles-catalyzed luminol?H2O2 chemiluminescence.
Khajvand T, et al.
Food Chemistry, 173, 514-520 (2015)
Multiresidue analysis of pesticides in fresh fruits and vegetables by supercritical fluid extraction and HPLC.
Kaihara A, et al.
Journal of Health Science, 46(5), 336-342 (2000)
Ana Masiá et al.
Journal of chromatography. A, 1345, 86-97 (2014-05-06)
Ultra-high pressure liquid chromatography-quadrupole time-of-flight mass spectrometry (UHPLC-QqTOF-MS) acquiring full scan MS data for quantification, and automatic data dependent information product ion spectra (IDA-MS/MS) without any predefinition of the ions by the user was checked for identifying organic contaminants in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service