Skip to Content
Merck
All Photos(2)

Key Documents

32996

Supelco

Sulfadimethoxine-d6

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€305.00

€305.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€305.00

About This Item

Empirical Formula (Hill Notation):
C12D6H8N4O4S
CAS Number:
Molecular Weight:
316.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€305.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+6

SMILES string

[2H]C([2H])([2H])Oc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC([2H])([2H])[2H])n1

InChI

1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)/i1D3,2D3

InChI key

ZZORFUFYDOWNEF-WFGJKAKNSA-N

General description

Sulfadimethoxine-d6 is an isotopically labeled compound of sulfonamide antibiotics. It is typically utilized as an analytical standard in chromatographic determinations.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfadoxine-d6 may be used as an analytical standard for the determination of various sulfonamides in:
  • Wastewater samples by ultra-performance liquid chromatography-triple quadrupole-linear ion trap mass spectrometer (UPLC-QqLit-MS).[1]
  • Fish samples[2] as well as soil, sludge and manure samples[3] by LC with tandem mass spectrometry (MS/MS).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue antimicrobial determination in Nile tilapia (Oreochromis Niloticus) cage farming by liquid chromatography tandem mass spectrometry.
Monteiro SH, et al.
Aquaculture (Amsterdam, Netherlands), 447, 37-43 (2015)
Evan Trofimchuk et al.
Journal of food science, 85(3), 736-743 (2020-02-06)
By utilizing the coffee-ring effect and microfluidic paper-based analytical devices (µPADs), this study improved the sensitivity of the determination of norfloxacin in four different food matrices. Micro-PADs in this study were fabricated by designing and embedding wax channels onto cellulose-based
Simultaneous extraction of four classes of antibiotics in soil, manure and sewage sludge and analysis by liquid chromatography-tandem mass spectrometry with the isotope-labelled internal standard method.
Huang Y, et al.
Analytical Methods : Advancing Methods and Applications, 5(15), 3721-3731 (2013)
Laura Micheli et al.
Nutrients, 12(6) (2020-06-04)
Current pharmacological therapies for the management of chronic articular diseases are far from being satisfactory, so new strategies need to be investigated. We tested the intra-articular pain relieving properties of a system of molecules from a characterized Centella asiatica extract
Run Tian et al.
Environmental pollution (Barking, Essex : 1987), 247, 1134-1142 (2019-03-03)
Antibiotics are introduced into agricultural fields by the application of manure or biosolids, or via irrigation using reclaimed wastewater. Antibiotics can enter the terrestrial food chains through plant uptake, which forms an alternative pathway for human exposure to antibiotics. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service