Skip to Content
Merck
All Photos(2)

Key Documents

32839

Supelco

Flubendazole-d3

VETRANAL®, analytical standard

Synonym(s):

Methyl-d3 [5-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D3H9FN3O3
CAS Number:
Molecular Weight:
316.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(F)cc3

InChI

1S/C16H12FN3O3/c1-23-16(22)20-15-18-12-7-4-10(8-13(12)19-15)14(21)9-2-5-11(17)6-3-9/h2-8H,1H3,(H2,18,19,20,22)/i1D3

InChI key

CPEUVMUXAHMANV-FIBGUPNXSA-N

General description

Flubendazole-d3 is an isotopically labeled compound of benzimidazole derivative, flubendazole.[1]

Application

Flubendazole-d3 may be used as an internal standard for the determination of flubendazole in animal muscle tissues[1] and sludge samples[2] by ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue method for identification and quantification of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue by ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) using a QuEChERS approach
Silva DRG, et al.
Talanta, 171, 307-320 (2017)
Damien Mooney et al.
Molecules (Basel, Switzerland), 24(10) (2019-05-28)
A comprehensive multiresidue method was developed and validated for the determination of 40 anthelmintic compounds, including 13 transformation products, in surface and groundwater samples at sub nanogram per litre (ng L-1) levels. Anthelmintic residues were extracted from unfiltered water samples
Xiaowei Li et al.
Journal of chromatography. A, 1499, 57-64 (2017-04-15)
This work reports the development of a multi-residue method for the identification and quantification of 82 veterinary drugs belonging to different chemical classes in swine waste lagoon. The proposed method applies a solid-phase extraction procedure with Oasis PRiME HLB cartridges

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service