Skip to Content
Merck
All Photos(2)

Key Documents

32828

Supelco

Clenproperol-d7

VETRANAL®, analytical standard

Synonym(s):

1-(4-Amino-3,5-dichlorophenyl)-2-isopropyl-d7-aminoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D7H9Cl2N2O
CAS Number:
Molecular Weight:
270.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+7

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])(NCC(O)c1cc(Cl)c(N)c(Cl)c1)C([2H])([2H])[2H]

InChI

1S/C11H16Cl2N2O/c1-6(2)15-5-10(16)7-3-8(12)11(14)9(13)4-7/h3-4,6,10,15-16H,5,14H2,1-2H3/i1D3,2D3,6D

InChI key

JXUDZCJTCKCTQK-NWOXSKRJSA-N

General description

Clenproperol-d7 is an isotopically labeled compound of the β-agonist, clenproperol.[1]

Application

Clenproperol-d7 may be used as an internal standard for the determination of clenproperol in biological samples[1][2][3] by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a multi-residue method for determination of 18β-agonists in bovine urine by UPLC-MS/MS.
Mauro D, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 115(1-2), 70-77 (2014)
Pseudo template molecularly imprinted polymer for determination of 14 kind of β-agonists in animal urine by ultra-high-performance liquid chromatography-tandem mass spectrometry.
Suo D, et al.
Journal of Chromatography A, 1526(1-2), 23-30 (2017)
The analysis of beta-agonists in bovine muscle using molecular imprinted polymers with ion trap LCMS screening.
Kootstra PR, et al.
Analytica Chimica Acta, 529(1-2), 75-81 (2005)
Suo Decheng et al.
Journal of chromatography. A, 1612, 460624-460624 (2019-11-02)
β-agonists, which have been illegally used in animal production in some countries, can induce bioaccumulation when blood is converted by rendering into blood meal. Unfortunately, available data on this topic are scarce, which result in lack of risk assessment. Therefore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service