Skip to Content
Merck
All Photos(2)

Key Documents

32569

Supelco

Cimaterol-d7

VETRANAL®, analytical standard

Synonym(s):

2-Amino-5-(1-hydroxy-2-isopropyl-d7-aminoethyl)benzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€815.00

€815.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€815.00

About This Item

Empirical Formula (Hill Notation):
C12D7H10N3O
CAS Number:
Molecular Weight:
226.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€815.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

mass shift

M+7

SMILES string

[2H]C([2H])([2H])C([2H])(NCC(O)c1ccc(N)c(c1)C#N)C([2H])([2H])[2H]

InChI

1S/C12H17N3O/c1-8(2)15-7-12(16)9-3-4-11(14)10(5-9)6-13/h3-5,8,12,15-16H,7,14H2,1-2H3/i1D3,2D3,8D

InChI key

BUXRLJCGHZZYNE-UNAVHCQLSA-N

Application

Cimaterol-d7 may be used as an internal standard for the determination of cimaterol in:
  • Bovine urine samples by ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS).[1]
  • Bovine muscles by molecular imprinted polymers (MIPs) with ion trap LC-MS.[2]

Animal muscles and viscera samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and LC-MS/MS.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of multiresidue analysis of ?-agonists in muscle and viscera using liquid chromatograph/tandem mass spectrometry with Quick, Easy, Cheap, Effective, Rugged, and Safe methodologies.
Lin YP, et al.
Journal of food and drug analysis, 25(2), 275-284 (2017)
Development and validation of a multi-residue method for determination of 18 ?-agonists in bovine urine by UPLC-MS/MS.
Mauro D, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 115, 70-77 (2014)
Yen-Ping Lin et al.
Journal of food and drug analysis, 25(2), 275-284 (2017-09-16)
The official analytical method of the Taiwan Food and Drug Administration, Ministry of Health and Welfare for testing for veterinary drug residues in foods is the multiresidue analysis of β-agonists. Samples are pretreated through liquid-liquid extraction and solid-phase extraction. This
The analysis of beta-agonists in bovine muscle using molecular imprinted polymers with ion trap LCMS screening.
Kootstra PR, et al.
Analytica Chimica Acta, 529(1-2), 75-81 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service